siRNA Details
Gene Symbol: LOC102554586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759935.1 | XP_008758157.1 | 3 | 333 | ||
GenBank mRNA | BG372695.1 | - | 1 | 363 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102554586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759935.1 | XP_008758157.1 | 3 | 333 | ||
GenBank mRNA | BG372695.1 | - | 1 | 363 |