siRNA Details
| Gene Symbol: Bag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011936.1 | NP_001011936.1 | 1, 2 | 288 | ||
| GenBank mRNA | BC085857.1 | - | 1, 2 | 288 | 7190249 | |
| DQ631552.1 | - | 1, 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bag3| Gene Symbol: Bag3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011936.1 | NP_001011936.1 | 1, 2 | 288 | ||
| GenBank mRNA | BC085857.1 | - | 1, 2 | 288 | 7190249 | |
| DQ631552.1 | - | 1, 2 | 291 | |||