siRNA Details
| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106323.1 | NP_001099793.1 | 5 | 696 | ||
| GenBank mRNA | FQ219373.1 | - | 5 | 645 | ||
| FQ229623.1 | - | 5 | 645 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhod| Gene Symbol: Rhod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106323.1 | NP_001099793.1 | 5 | 696 | ||
| GenBank mRNA | FQ219373.1 | - | 5 | 645 | ||
| FQ229623.1 | - | 5 | 645 | |||