siRNA Details
| Gene Symbol: Fgf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019198.1 | NP_062071.1 | 4 | 312 | ||
| XM_008770827.1 | XP_008769049.1 | 5 | 468 | |||
| GenBank mRNA | AB008682.1 | - | 4 | 312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf17| Gene Symbol: Fgf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019198.1 | NP_062071.1 | 4 | 312 | ||
| XM_008770827.1 | XP_008769049.1 | 5 | 468 | |||
| GenBank mRNA | AB008682.1 | - | 4 | 312 | ||