siRNA Details
| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106331.1 | NP_001099801.1 | 4 | 727 | ||
| XM_006230761.2 | XP_006230823.1 | 4 | 417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpt1| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106331.1 | NP_001099801.1 | 4 | 727 | ||
| XM_006230761.2 | XP_006230823.1 | 4 | 417 | |||