siRNA Details
| Gene Symbol: Glyat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009648.2 | NP_001009648.1 | 5 | 724 | ||
| XM_006231121.2 | XP_006231183.1 | 7 | 1031 | |||
| XM_006231122.2 | XP_006231184.1 | 6 | 946 | |||
| XM_006231123.2 | XP_006231185.1 | 6 | 768 | |||
| XM_008760257.1 | XP_008758479.1 | 7 | 1911 | |||
| GenBank mRNA | AB201944.1 | - | 5 | 724 | ||
| AY325192.1 | - | 7 | 911 | |||
| BC087043.1 | - | 6 | 822 | 7108337 | ||

Glyat
Genomic Map