siRNA Details
| Gene Symbol: Tpd52l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106349.1 | NP_001099819.1 | 1 | 341 | ||
| XM_008760324.1 | XP_008758546.1 | 1 | 1380 | |||
| Gene Symbol: LOC103690131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231260.2 | XP_006231322.1 | 1380 | |||
| XM_008760353.1 | XP_008758575.1 | 1379 | ||||

Tpd52l3
Genomic Map