siRNA Details
Gene Symbol: Tpd52l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106349.1 | NP_001099819.1 | 1 | 33 | ||
XM_008760324.1 | XP_008758546.1 | 1 | 1072 |
Gene Symbol: LOC103690131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231260.2 | XP_006231322.1 | 1072 | |||
XM_008760353.1 | XP_008758575.1 | 1071 |
