siRNA Details
| Gene Symbol: Zfand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106356.1 | NP_001099826.1 | 1 | 144 | ||
| XM_008760297.1 | XP_008758519.1 | 2 | 726 | |||
| GenBank mRNA | BC168882.1 | - | 2 | 194 | 9026652 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfand5| Gene Symbol: Zfand5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106356.1 | NP_001099826.1 | 1 | 144 | ||
| XM_008760297.1 | XP_008758519.1 | 2 | 726 | |||
| GenBank mRNA | BC168882.1 | - | 2 | 194 | 9026652 | |