siRNA Details
| Gene Symbol: Adra1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024483.1 | NP_077809.1 | 1 | 501 | ||
| GenBank mRNA | L31771.1 | - | 1 | 501 | ||
| M60654.1 | - | 1 | 501 | |||
More Information
Other Species Targeted
Set Membership: |
|

Adra1d
Genomic Map