siRNA Details
| Gene Symbol: Atn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | U31777.1 | - | 5 | 1472 | ||
| Gene Symbol: LOC100911672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750658.3 | XP_003750706.1 | 5 | 1459 | ||
| XM_006244637.2 | XP_006244699.1 | 5 | 1445 | |||
| XM_008766913.1 | XP_008765135.1 | 5 | 1427 | |||

Atn1
Genomic Map