siRNA Details
| Gene Symbol: Rbm8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271138.1 | NP_001258067.1 | 4 | 308 | ||
| XM_006232956.2 | XP_006233018.1 | 4 | 306 | |||
| GenBank mRNA | DQ359102.1 | - | 4 | 260 | ||
| FQ210627.1 | - | 4 | 281 | |||

Rbm8a
Genomic Map