siRNA Details
| Gene Symbol: Tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007643.1 | NP_001007644.1 | 9 | 1121 | ||
| GenBank mRNA | BC083678.1 | - | 9 | 1121 | 7135830 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmx2| Gene Symbol: Tmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007643.1 | NP_001007644.1 | 9 | 1121 | ||
| GenBank mRNA | BC083678.1 | - | 9 | 1121 | 7135830 | |