siRNA Details
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017252.2 | NP_058948.1 | 1 | 55 | ||
GenBank mRNA | M84645.1 | - | 1 | 55 | ||
Z11834.1 | - | 1 | 27 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pou3f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017252.2 | NP_058948.1 | 1 | 55 | ||
GenBank mRNA | M84645.1 | - | 1 | 55 | ||
Z11834.1 | - | 1 | 27 |