siRNA Details
| Gene Symbol: Pou3f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017252.2 | NP_058948.1 | 1 | 586 | ||
| GenBank mRNA | M84645.1 | - | 1 | 586 | ||
| Z11834.1 | - | 1 | 558 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou3f4| Gene Symbol: Pou3f4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017252.2 | NP_058948.1 | 1 | 586 | ||
| GenBank mRNA | M84645.1 | - | 1 | 586 | ||
| Z11834.1 | - | 1 | 558 | |||