siRNA Details
| Gene Symbol: Cacng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019255.1 | NP_062128.1 | 2, 3 | 315 | ||
| GenBank mRNA | FQ215004.1 | - | 2, 3 | 412 | ||
| FQ215168.1 | - | 2, 3 | 394 | |||
| FQ215755.1 | - | 2, 3 | 394 | |||
| FQ216096.1 | - | 2, 3 | 430 | |||
| FQ216734.1 | - | 2, 3 | 430 | |||
| FQ224130.1 | - | 2, 3 | 486 | |||
| FQ224197.1 | - | 2, 3 | 409 | |||
| Y09453.1 | - | 2, 3 | 315 | |||

Cacng1
Genomic Map