siRNA Details
| Gene Symbol: Phyhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013081.1 | NP_001013099.1 | 6 | 459 | ||
| XM_006233777.2 | XP_006233839.1 | 7 | 843 | |||
| XM_006233778.2 | XP_006233840.1 | 7 | 843 | |||
| XM_006233779.2 | XP_006233841.1 | 7 | 1536 | |||
| XM_006233780.2 | XP_006233842.1 | 7 | 599 | |||
| GenBank mRNA | BC091389.1 | - | 6 | 459 | 7321523 | |
| FQ212963.1 | - | 6 | 475 | |||
| FQ219402.1 | - | 6 | 480 | |||
| FQ219572.1 | - | 3 | 462 | |||
| FQ223356.1 | - | 6 | 466 | |||

Phyhd1
Genomic Map