siRNA Details
| Gene Symbol: Olr416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000387.1 | NP_001000387.1 | 1 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr416| Gene Symbol: Olr416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000387.1 | NP_001000387.1 | 1 | 70 | ||