siRNA Details
| Gene Symbol: Mrpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029898.1 | NP_001025069.1 | 5 | 736 | ||
| GenBank mRNA | FQ235078.1 | - | 5 | 754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl19| Gene Symbol: Mrpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029898.1 | NP_001025069.1 | 5 | 736 | ||
| GenBank mRNA | FQ235078.1 | - | 5 | 754 | ||