siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750036.3 | XP_003750084.3 | 4, 5 | 766 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb8os| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750036.3 | XP_003750084.3 | 4, 5 | 766 | ||