siRNA Details
Gene Symbol: Nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207596.1 | NP_997479.1 | 3 | 231 | ||
GenBank mRNA | BC058156.1 | - | 3 | 231 | 6888562 | |
FQ227408.1 | - | 3 | 232 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207596.1 | NP_997479.1 | 3 | 231 | ||
GenBank mRNA | BC058156.1 | - | 3 | 231 | 6888562 | |
FQ227408.1 | - | 3 | 232 |