siRNA Details
| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106659.1 | NP_001100129.1 | 14 | 1558 | ||
| GenBank mRNA | BC087165.1 | - | 13 | 1615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf4| Gene Symbol: Prpf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106659.1 | NP_001100129.1 | 14 | 1558 | ||
| GenBank mRNA | BC087165.1 | - | 13 | 1615 | ||