siRNA Details
| Gene Symbol: Rbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034135.1 | NP_001029307.1 | 115 | |||
| GenBank mRNA | BC100258.1 | - | 115 | 7444286 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbx1| Gene Symbol: Rbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034135.1 | NP_001029307.1 | 115 | |||
| GenBank mRNA | BC100258.1 | - | 115 | 7444286 | ||