siRNA Details
| Gene Symbol: Pfdn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106794.2 | NP_001100264.2 | 5, 6 | 429 | ||
| GenBank mRNA | AI576681.1 | - | 1, 2 | 228 | ||
| BC169128.1 | - | 5, 6 | 395 | 6921287 | ||
| CF976389.1 | - | 5, 6 | 433 | |||
| CF976393.1 | - | 5, 6 | 400 | |||

Pfdn5
Genomic Map