siRNA Details
| Gene Symbol: Pfdn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106794.2 | NP_001100264.2 | 4 | 281 | ||
| GenBank mRNA | BC169128.1 | - | 4 | 247 | 6921287 | |
| CF976389.1 | - | 4 | 285 | |||
| CF976393.1 | - | 4 | 252 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pfdn5| Gene Symbol: Pfdn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106794.2 | NP_001100264.2 | 4 | 281 | ||
| GenBank mRNA | BC169128.1 | - | 4 | 247 | 6921287 | |
| CF976389.1 | - | 4 | 285 | |||
| CF976393.1 | - | 4 | 252 | |||