siRNA Details
| Gene Symbol: Olr1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000462.1 | NP_001000462.1 | 844 | |||
| XM_003750500.1 | XP_003750548.1 | 1 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1301| Gene Symbol: Olr1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000462.1 | NP_001000462.1 | 844 | |||
| XM_003750500.1 | XP_003750548.1 | 1 | 406 | |||