siRNA Details
| Gene Symbol: Spats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181376.2 | NP_852041.1 | 6, 7 | 765 | ||
| GenBank mRNA | AY157978.1 | - | 6, 7 | 765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spats1| Gene Symbol: Spats1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181376.2 | NP_852041.1 | 6, 7 | 765 | ||
| GenBank mRNA | AY157978.1 | - | 6, 7 | 765 | ||