siRNA Details
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009674.1 | NP_001009674.1 | 3 | 377 | ||
GenBank mRNA | BC087127.1 | - | 3 | 377 | 7111761 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009674.1 | NP_001009674.1 | 3 | 377 | ||
GenBank mRNA | BC087127.1 | - | 3 | 377 | 7111761 |