siRNA Details
| Gene Symbol: Pdzd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106945.1 | NP_001100415.1 | 6, 7 | 576 | ||
| XM_006257068.2 | XP_006257130.1 | 6, 7 | 482 | |||
| XM_006257070.2 | XP_006257132.1 | 5, 6 | 417 | |||
| GenBank mRNA | BC168740.1 | - | 5, 6 | 340 | 6918646 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_028303, AK012624, AK032327, AK007454, BC004608.

Pdzd11
Genomic Map