siRNA Details
| Gene Symbol: Pdzd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106945.1 | NP_001100415.1 | 2 | 226 | ||
| XM_006257068.2 | XP_006257130.1 | 2 | 132 | |||
| XM_006257070.2 | XP_006257132.1 | 2 | 151 | |||
| GenBank mRNA | BC168740.1 | - | 2 | 74 | 6918646 | |

Pdzd11
Genomic Map