siRNA Details
| Gene Symbol: MGC114483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024880.1 | NP_001020051.1 | 3 | 830 | ||
| XM_006257008.2 | XP_006257070.1 | 4 | 1181 | |||
| XM_006257009.2 | XP_006257071.1 | 4 | 901 | |||
| XM_006257010.2 | XP_006257072.1 | 3 | 814 | |||
| GenBank mRNA | BC097433.1 | - | 3 | 830 | 7455406 | |

MGC114483
Genomic Map