siRNA Details
Gene Symbol: Pqbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013957.1 | NP_001013979.1 | 4 | 400 | ||
XM_006256707.1 | XP_006256769.1 | 4 | 374 | |||
XM_006256708.2 | XP_006256770.1 | 3 | 275 | |||
GenBank mRNA | BC059163.1 | - | 4 | 400 | 6889015 |
