siRNA Details
| Gene Symbol: Syngr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106985.1 | NP_001100455.1 | 3 | 495 | ||
| XM_006245947.2 | XP_006246009.2 | 3 | 495 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syngr3| Gene Symbol: Syngr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106985.1 | NP_001100455.1 | 3 | 495 | ||
| XM_006245947.2 | XP_006246009.2 | 3 | 495 | |||