siRNA Details
| Gene Symbol: LOC103689949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006246253.2 | XP_006246315.1 | 3 | 954 | ||
| Gene Symbol: N4bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033893.1 | NP_001029065.1 | 2 | 731 | ||
| XM_008767681.1 | XP_008765903.1 | 954 | ||||
| GenBank mRNA | DQ176639.1 | - | 2 | 731 | ||

LOC103689949
Genomic Map