siRNA Details
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107011.1 | NP_001100481.1 | 6, 7 | 655 | ||
GenBank mRNA | BC087071.1 | - | 6, 7 | 832 | ||
BC107940.1 | - | 4, 5 | 861 | |||
FQ213141.1 | - | 6, 7 | 631 | |||
FQ220130.1 | - | 6, 7 | 619 | |||
FQ229625.1 | - | 4, 5 | 424 | |||
FQ229903.1 | - | 4, 5 | 424 |
