siRNA Details
Gene Symbol: Mpdu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107011.1 | NP_001100481.1 | 4 | 358 | ||
GenBank mRNA | BC087071.1 | - | 4 | 535 | ||
BC107940.1 | - | 4 | 317 | |||
FQ213141.1 | - | 4 | 334 | |||
FQ220130.1 | - | 4 | 322 | |||
FQ229625.1 | - | 2 | 127 | |||
FQ229903.1 | - | 2 | 127 |
