siRNA Details
| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009369.2 | NP_001009369.2 | 5 | 633 | ||
| XM_008768372.1 | XP_008766594.1 | 5 | 1421 | |||
| GenBank mRNA | BC087096.1 | - | 5 | 630 | 7191805 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl38| Gene Symbol: Mrpl38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009369.2 | NP_001009369.2 | 5 | 633 | ||
| XM_008768372.1 | XP_008766594.1 | 5 | 1421 | |||
| GenBank mRNA | BC087096.1 | - | 5 | 630 | 7191805 | |