siRNA Details
| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107076.1 | NP_001100546.1 | 4 | 417 | ||
| GenBank mRNA | BC168956.1 | - | 4 | 332 | 7123800 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab40b| Gene Symbol: Rab40b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107076.1 | NP_001100546.1 | 4 | 417 | ||
| GenBank mRNA | BC168956.1 | - | 4 | 332 | 7123800 | |