siRNA Details
| Gene Symbol: Amz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047092.1 | NP_001040557.1 | 3 | 573 | ||
| XM_006248930.2 | XP_006248992.1 | 6 | 1109 | |||
| XM_006248931.2 | XP_006248993.1 | 5 | 996 | |||
| XM_006248932.2 | XP_006248994.1 | 4 | 1406 | |||
| XM_006248933.2 | XP_006248995.1 | 4 | 1323 | |||
| XM_008768970.1 | XP_008767192.1 | 4 | 1111 | |||
| GenBank mRNA | AJ879913.1 | - | 3 | 573 | ||
| BC160815.1 | - | 3 | 592 | 7365526 | ||

Amz1
Genomic Map