siRNA Details
| Gene Symbol: Tnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024771.1 | NP_001019942.1 | 2 | 469 | ||
| XM_006251255.2 | XP_006251317.1 | 2 | 816 | |||
| GenBank mRNA | BC085912.1 | - | 2 | 400 | ||
| BC097956.1 | - | 2 | 469 | 7461734 | ||

Tnip2
Genomic Map