siRNA Details
| Gene Symbol: Gpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001077762.5 | XP_001077762.3 | 9 | 2536 | ||
| XM_003751533.3 | XP_003751581.1 | 10 | 2567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc6| Gene Symbol: Gpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001077762.5 | XP_001077762.3 | 9 | 2536 | ||
| XM_003751533.3 | XP_003751581.1 | 10 | 2567 | |||