siRNA Details
| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107338.1 | NP_001100808.1 | 5 | 681 | ||
| XM_006253628.2 | XP_006253690.1 | 5 | 734 | |||
| GenBank mRNA | FQ212045.1 | - | 5 | 725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp346| Gene Symbol: Zfp346 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107338.1 | NP_001100808.1 | 5 | 681 | ||
| XM_006253628.2 | XP_006253690.1 | 5 | 734 | |||
| GenBank mRNA | FQ212045.1 | - | 5 | 725 | ||