siRNA Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004256.1 | NP_001004256.1 | 4 | 574 | ||
| XM_006253801.1 | XP_006253863.1 | 5 | 758 | |||
| XM_006253802.1 | XP_006253864.1 | 5 | 636 | |||
| XM_006253803.2 | XP_006253865.1 | 5 | 630 | |||
| XM_006253804.2 | XP_006253866.1 | 5 | 581 | |||
| GenBank mRNA | BC078958.1 | - | 4 | 574 | 7110841 | |

Sirt5
Genomic Map