siRNA Details
| Gene Symbol: Gcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001511.2 | NP_001001511.1 | 1 | 669 | ||
| GenBank mRNA | AY435153.1 | - | 1 | 189 | ||
| BC099796.1 | - | 1 | 669 | 7936188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gcnt2| Gene Symbol: Gcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001511.2 | NP_001001511.1 | 1 | 669 | ||
| GenBank mRNA | AY435153.1 | - | 1 | 189 | ||
| BC099796.1 | - | 1 | 669 | 7936188 | ||