siRNA Details
| Gene Symbol: Fam50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107349.1 | NP_001100819.1 | 2 | 541 | ||
| XM_006253827.2 | XP_006253889.1 | 2 | 895 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam50b| Gene Symbol: Fam50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107349.1 | NP_001100819.1 | 2 | 541 | ||
| XM_006253827.2 | XP_006253889.1 | 2 | 895 | |||