siRNA Details
Gene Symbol: Mesdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008345.1 | NP_001008346.1 | 1 | 288 | ||
GenBank mRNA | BC085892.1 | - | 1 | 288 | 7192668 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mesdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008345.1 | NP_001008346.1 | 1 | 288 | ||
GenBank mRNA | BC085892.1 | - | 1 | 288 | 7192668 |