siRNA Details
| Gene Symbol: Rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015012.1 | NP_001015012.1 | 2 | 246 | ||
| XM_006229692.2 | XP_006229754.1 | 4 | 471 | |||
| XM_006229693.2 | XP_006229755.1 | 4 | 440 | |||
| XM_008759647.1 | XP_008757869.1 | 3 | 3438 | |||
| GenBank mRNA | BC091182.1 | - | 2 | 246 | 7326213 | |
| FQ210359.1 | - | 3 | 347 | |||

Rab30
Genomic Map