siRNA Details
| Gene Symbol: Rhog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037195.1 | NP_001032272.1 | 2 | 149 | ||
| XM_006229882.2 | XP_006229944.1 | 3 | 290 | |||
| XM_006229883.2 | XP_006229945.1 | 2 | 396 | |||
| GenBank mRNA | BC107943.1 | - | 2 | 149 | 7389075 | |
| FQ212590.1 | - | 2 | 189 | |||
| FQ232547.1 | - | 2 | 192 | |||

Rhog
Genomic Map