siRNA Details
| Gene Symbol: Aldh3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006998.1 | NP_001006999.1 | 3 | 280 | ||
| XM_006230787.2 | XP_006230849.1 | 4 | 583 | |||
| XM_008760132.1 | XP_008758354.1 | 3 | 770 | |||
| XM_008760133.1 | XP_008758355.1 | 3 | 770 | |||
| XM_008760134.1 | XP_008758356.1 | 3 | 264 | |||
| GenBank mRNA | BC083850.1 | - | 3 | 280 | 7122451 | |

Aldh3b1
Genomic Map