siRNA Details
| Gene Symbol: Adh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012084.1 | NP_001012084.1 | 7 | 998 | ||
| XM_006233372.2 | XP_006233434.1 | 7 | 1792 | |||
| XM_006233374.1 | XP_006233436.1 | 8 | 1070 | |||
| XM_006233375.2 | XP_006233437.1 | 9 | 1281 | |||
| XM_006233377.2 | XP_006233439.1 | 7 | 1240 | |||
| XM_006233378.2 | XP_006233440.1 | 8 | 1150 | |||
| XM_006233379.2 | XP_006233441.1 | 8 | 1147 | |||
| XM_008761506.1 | XP_008759728.1 | 6 | 1112 | |||
| XM_008761507.1 | XP_008759729.1 | 8 | 3344 | |||
| XM_008761508.1 | XP_008759730.1 | 9 | 3204 | |||
| XM_008761509.1 | XP_008759731.1 | 9 | 1696 | |||
| XM_008761510.1 | XP_008759732.1 | 9 | 3472 | |||
| GenBank mRNA | BC083792.1 | - | 7 | 998 | 7106499 | |

Adh6
Genomic Map